Assay Details
| Gene Symbol: SHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075305.2 | NP_001068773.1 | 3 - 4 | 325 | 58 | |
| NM_001164061.1 | NP_001157533.1 | 2 - 3 | 764 | 58 | ||
| GenBank mRNA | BC122688.1 | - | 3 - 4 | 312 | 58 | |
| DQ125497.1 | - | 2 - 3 | 240 | 58 | ||
| EH179100.1 | - | 4 - 5 | 474 | 58 | ||
More Information
Set Membership: |
|

SHC1
Genomic Map