Assay Details
| Gene Symbol: IP6K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076010.1 | NP_001069478.1 | 4 - 5 | 956 | 59 | |
| GenBank mRNA | BC122746.1 | - | 4 - 5 | 956 | 59 | |
More Information
Set Membership: |
|

IP6K1
Genomic Map