Assay Details
| Gene Symbol: ANKRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046289.2 | NP_001039754.1 | 2 - 3 | 716 | 66 | |
| GenBank mRNA | BC112737.1 | - | 2 - 3 | 717 | 66 | |
More Information
Set Membership: |
|

ANKRA2
Genomic Map