Assay Details
| Gene Symbol: RSPO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076034.1 | NP_001069502.1 | 2 - 3 | 630 | 155 | |
| GenBank mRNA | BC114713.1 | - | 2 - 3 | 630 | 155 | |
More Information
Set Membership: |
|

RSPO3
Genomic Map