Assay Details
| Gene Symbol: COP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103256.1 | NP_001096726.1 | 13 - 14 | 1630 | 95 | |
| GenBank mRNA | BC133612.1 | - | 13 - 14 | 1630 | 95 | |
More Information
Set Membership: |
|

COP1
Genomic Map