Assay Details
| Gene Symbol: UBAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100376.1 | NP_001093846.1 | 3 - 4 | 328 | 78 | |
| GenBank mRNA | BC114036.1 | - | 4 - 5 | 470 | 78 | |
| BC150086.1 | - | 3 - 4 | 328 | 78 | ||
More Information
Set Membership: |
|

UBAP1
Genomic Map