Assay Details
| Gene Symbol: UBA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075574.1 | NP_001069042.1 | 2 - 3 | 70 | 73 | |
| GenBank mRNA | BC119988.1 | - | 2 - 3 | 70 | 73 | |
More Information
Set Membership: |
|

UBA3
Genomic Map