Assay Details
| Gene Symbol: PSMD14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076067.1 | NP_001069535.1 | 11 - 12 | 1125 | 66 | |
| GenBank mRNA | BC118241.1 | - | 11 - 12 | 1125 | 66 | |
More Information
Set Membership: |
|

PSMD14
Genomic Map