Assay Details
| Gene Symbol: YTHDF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076253.1 | NP_001069721.1 | 2 - 3 | 224 | 118 | |
| GenBank mRNA | BC119920.1 | - | 2 - 3 | 224 | 118 | |
More Information
Set Membership: |
|

YTHDF2
Genomic Map