Assay Details
| Gene Symbol: YWHAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174814.2 | NP_777239.1 | 2 - 3 | 345 | 120 | |
| GenBank mRNA | BC102382.1 | - | 2 - 3 | 379 | 120 | |
| L07955.1 | - | 2 - 3 | 345 | 120 | ||
More Information
Set Membership: |
|

YWHAZ
Genomic Map