Assay Details
| Gene Symbol: DAZL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081725.1 | NP_001075194.1 | 4 - 5 | 446 | 71 | |
| GenBank mRNA | DQ408764.1 | - | 4 - 5 | 446 | 71 | |
| DQ408765.1 | - | 4 - 5 | 446 | 71 | ||
| EF501823.2 | - | 4 - 5 | 464 | 71 | ||
More Information
Set Membership: |
|

DAZL
Genomic Map