Assay Details
| Gene Symbol: IPMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035462.1 | NP_001030539.1 | 1 - 2 | 497 | 69 | |
| GenBank mRNA | BC102793.1 | - | 1 - 2 | 497 | 69 | |
More Information
Set Membership: |
|

IPMK
Genomic Map