Assay Details
| Gene Symbol: SZRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114608.2 | NP_001108080.1 | 2 - 3 | 128 | 83 | |
| GenBank mRNA | BE666569.1 | - | 2 - 3 | 89 | 83 | |
| EE237864.1 | - | 2 - 3 | 121 | 83 | ||
| EH204951.1 | - | 2 - 3 | 119 | 83 | ||
More Information
Set Membership: |
|

SZRD1
Genomic Map