Assay Details
| Gene Symbol: KIF3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078155.1 | NP_001071623.1 | 3 - 4 | 2130 | 69 | |
| GenBank mRNA | BC126505.1 | - | 3 - 4 | 2130 | 69 | |
More Information
Set Membership: |
|

KIF3C
Genomic Map