Assay Details
| Gene Symbol: CLPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046414.2 | NP_001039879.1 | 1 - 2 | 233 | 68 | |
| GenBank mRNA | BC112880.1 | - | 1 - 2 | 234 | 68 | |
More Information
Set Membership: |
|

CLPP
Genomic Map