Assay Details
| Gene Symbol: ALDOB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034485.2 | NP_001029657.1 | 7 - 8 | 879 | 67 | |
| GenBank mRNA | BC102278.1 | - | 7 - 8 | 880 | 67 | |
| BC142385.1 | - | 7 - 8 | 988 | 67 | ||
More Information
Set Membership: |
|

ALDOB
Genomic Map