Assay Details
| Gene Symbol: WASF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034017.3 | NP_001029189.2 | 6 - 7 | 1145 | 79 | |
| GenBank mRNA | BC122612.1 | - | 6 - 7 | 912 | 79 | |
More Information
Set Membership: |
|

WASF1
Genomic Map