Assay Details
| Gene Symbol: DPY30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077916.2 | NP_001071384.1 | 4 - 5 | 330 | 124 | |
| GenBank mRNA | BC111634.1 | - | 4 - 5 | 333 | 124 | |
More Information
Set Membership: |
|

DPY30
Genomic Map