Assay Details
| Gene Symbol: NAP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099215.2 | NP_001092685.1 | 9 - 10 | 952 | 143 | |
| GenBank mRNA | BC146131.1 | - | 9 - 10 | 953 | 143 | |
More Information
Set Membership: |
|

NAP1L1
Genomic Map