Assay Details
| Gene Symbol: PPP2CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181031.2 | NP_851374.1 | 4 - 5 | 705 | 70 | |
| GenBank mRNA | BC147979.1 | - | 4 - 5 | 794 | 70 | |
| BT021727.1 | - | 4 - 5 | 584 | 70 | ||
| M16968.1 | - | 4 - 5 | 683 | 70 | ||
| X52554.1 | - | 705 | 70 | |||
| X72858.1 | - | 743 | 70 | |||
More Information
Set Membership: |
|

PPP2CA
Genomic Map