Assay Details
| Gene Symbol: PPP1CC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174581.3 | NP_777006.1 | 1 - 2 | 278 | 76 | |
| GenBank mRNA | AJ429235.1 | - | 1 - 2 | 77 | 76 | |
| BC123502.1 | - | 1 - 2 | 278 | 76 | ||
More Information
Set Membership: |
|

PPP1CC
Genomic Map