Assay Details
| Gene Symbol: U2AF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080268.2 | NP_001073737.1 | 2 - 3 | 171 | 83 | |
| GenBank mRNA | BC126638.1 | - | 2 - 3 | 172 | 83 | |
More Information
Set Membership: |
|

U2AF1
Genomic Map