Assay Details
| Gene Symbol: EIF4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174310.3 | NP_776735.2 | 6 - 7 | 559 | 145 | |
| GenBank mRNA | AF257235.1 | - | 6 - 7 | 558 | 145 | |
| BC111272.1 | - | 6 - 7 | 559 | 145 | ||
More Information
Set Membership: |
|

EIF4E
Genomic Map