Assay Details
| Gene Symbol: BOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076200.1 | NP_001069668.1 | 1 - 2 | 382 | 138 | |
| GenBank mRNA | BC114807.1 | - | 1 - 2 | 382 | 138 | |
More Information
Set Membership: |
|

BOD1
Genomic Map