Assay Details
| Gene Symbol: UBE3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098462.1 | NP_001091932.1 | 5 - 6 | 1774 | 113 | |
| GenBank mRNA | BC134678.1 | - | 5 - 6 | 1774 | 113 | |
More Information
Set Membership: |
|

UBE3A
Genomic Map