Assay Details
| Gene Symbol: DENR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046528.1 | NP_001039993.1 | 4 - 5 | 298 | 107 | |
| GenBank mRNA | BC113316.1 | - | 4 - 5 | 298 | 107 | |
More Information
Set Membership: |
|

DENR
Genomic Map