Assay Details
| Gene Symbol: SLC9A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174833.2 | NP_777258.1 | 9 - 10 | 2343 | 64 | |
| GenBank mRNA | U49432.1 | - | 9 - 10 | 2343 | 64 | |
More Information
Set Membership: |
|

SLC9A1
Genomic Map