Assay Details
| Gene Symbol: ACTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098052.1 | NP_001091521.1 | 7 - 8 | 774 | 100 | |
| GenBank mRNA | BC140512.1 | - | 7 - 8 | 774 | 100 | |
More Information
Set Membership: |
|

ACTN4
Genomic Map