Assay Details
| Gene Symbol: SYT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098115.1 | NP_001091584.1 | 2 - 3 | 484 | 82 | |
| GenBank mRNA | BC140518.1 | - | 2 - 3 | 484 | 82 | |
More Information
Set Membership: |
|

SYT13
Genomic Map