Assay Details
| Gene Symbol: SUPT20H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046370.1 | NP_001039835.1 | 11 - 12 | 1012 | 91 | |
| GenBank mRNA | BC105571.1 | - | 11 - 12 | 1012 | 91 | |
More Information
Set Membership: |
|

SUPT20H
Genomic Map