Assay Details
| Gene Symbol: PRLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039726.2 | NP_001034815.1 | 3 - 4 | 289 | 72 | |
| NM_174155.3 | NP_776580.1 | 3 - 4 | 289 | 72 | ||
| GenBank mRNA | AF027403.1 | - | 3 - 4 | 291 | 72 | |
| L02549.1 | - | 3 - 4 | 291 | 72 | ||
More Information
Set Membership: |
|

PRLR
Genomic Map