Assay Details
| Gene Symbol: MOGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001154.1 | NP_001001154.1 | 3 - 4 | 541 | 76 | |
| GenBank mRNA | AJ519786.1 | - | 3 - 4 | 541 | 76 | |
More Information
Set Membership: |
|

MOGAT2
Genomic Map