Assay Details
| Gene Symbol: MAP1LC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001169.1 | NP_001001169.1 | 3 - 4 | 245 | 112 | |
| GenBank mRNA | AY570553.1 | - | 3 - 4 | 245 | 112 | |
| BC102891.1 | - | 3 - 4 | 299 | 112 | ||
More Information
Set Membership: |
|

MAP1LC3B
Genomic Map