Assay Details
| Gene Symbol: ATG4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001171.1 | NP_001001171.1 | 6 - 7 | 546 | 69 | |
| GenBank mRNA | AY570549.1 | - | 6 - 7 | 546 | 69 | |
| BC147962.1 | - | 7 - 8 | 591 | 69 | ||
More Information
Set Membership: |
|

ATG4A
Genomic Map