Assay Details
| Gene Symbol: CDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002763.1 | NP_001002763.1 | 9 - 10 | 1315 | 60 | |
| GenBank mRNA | AY508164.1 | - | 9 - 10 | 1315 | 60 | |
| BC147914.1 | - | 9 - 10 | 1425 | 60 | ||
More Information
Set Membership: |
|

CDH1
Genomic Map