Assay Details
| Gene Symbol: COX5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002891.1 | NP_001002891.1 | 4 - 5 | 567 | 153 | |
| GenBank mRNA | AY528254.1 | - | 4 - 5 | 567 | 153 | |
More Information
Set Membership: |
|

COX5A
Genomic Map