Assay Details
| Gene Symbol: DYNLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003901.1 | NP_001003901.1 | 2 - 3 | 232 | 112 | |
| GenBank mRNA | AB098827.1 | - | 1 - 2 | 328 | 112 | |
| AB098977.1 | - | 2 - 3 | 220 | 112 | ||
| AB099086.1 | - | 2 - 3 | 220 | 112 | ||
| AY675078.1 | - | 2 - 3 | 232 | 112 | ||
| BC102872.1 | - | 2 - 3 | 233 | 112 | ||
| BT021030.1 | - | 2 - 3 | 278 | 112 | ||
More Information
Set Membership: |
|

DYNLL1
Genomic Map