Assay Details
| Gene Symbol: PTGES3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007806.2 | NP_001007807.2 | 7 - 8 | 656 | 88 | |
| GenBank mRNA | BC103350.1 | - | 7 - 8 | 656 | 88 | |
More Information
Set Membership: |
|

PTGES3
Genomic Map