Assay Details
| Gene Symbol: PNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007818.1 | NP_001007819.1 | 3 - 4 | 375 | 71 | |
| GenBank mRNA | AF529136.1 | - | 3 - 4 | 375 | 71 | |
| BC103291.1 | - | 3 - 4 | 372 | 71 | ||
| BT021545.1 | - | 3 - 4 | 371 | 71 | ||
More Information
Set Membership: |
|

PNP
Genomic Map