Assay Details
| Gene Symbol: KLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008416.1 | NP_001008416.1 | 3 - 4 | 532 | 135 | |
| GenBank mRNA | AY290705.1 | - | 3 - 4 | 532 | 135 | |
| BC151558.1 | - | 3 - 4 | 527 | 135 | ||
More Information
Set Membership: |
|

KLK1
Genomic Map