Assay Details
| Gene Symbol: KRT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008663.1 | NP_001008663.1 | 5 - 6 | 1097 | 69 | |
| GenBank mRNA | BC140616.1 | - | 5 - 6 | 1198 | 69 | |
| BC149167.1 | - | 3291 | 69 | |||
More Information
Set Membership: |
|

KRT5
Genomic Map