Assay Details
| Gene Symbol: CRABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008670.1 | NP_001008670.1 | 3 - 4 | 371 | 76 | |
| GenBank mRNA | AY821680.1 | - | 3 - 4 | 371 | 76 | |
| BC142037.1 | - | 3 - 4 | 498 | 76 | ||
More Information
Set Membership: |
|

CRABP2
Genomic Map