Assay Details
| Gene Symbol: CXCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011673.1 | NP_001011673.1 | 1 | 955 | 80 | |
| GenBank mRNA | AY834254.1 | - | 1 | 955 | 80 | |
| BT030690.1 | - | 2 | 1080 | 80 | ||
More Information
Set Membership: |
|

CXCR3
Genomic Map