Assay Details
| Gene Symbol: BOLA-DQA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012675.2 | NP_001012693.2 | 4 - 5 | 850 | 93 | |
| GenBank mRNA | AF037314.1 | - | 4 - 5 | 840 | 93 | |
| EV733097.1 | - | 1 - 2 | 335 | 93 | ||
More Information
Set Membership: |
|

BOLA-DQA5
Genomic Map