Assay Details
| Gene Symbol: BOLA-DYA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012678.1 | NP_001012696.1 | 2 - 3 | 433 | 112 | |
| GenBank mRNA | AJ580583.2 | - | 2 - 3 | 433 | 112 | |
| BC149795.1 | - | 2 - 3 | 369 | 112 | ||
More Information
Set Membership: |
|

BOLA-DYA
Genomic Map