Assay Details
| Gene Symbol: CRYGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013595.1 | NP_001013613.1 | 2 - 3 | 275 | 50 | |
| GenBank mRNA | EF426310.1 | - | 2 - 3 | 275 | 50 | |
| L27069.1 | - | 2 - 3 | 275 | 50 | ||
More Information
Set Membership: |
|

CRYGC
Genomic Map