Assay Details
| Gene Symbol: RPS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014387.2 | NP_001014409.1 | 1 - 2 | 58 | 80 | |
| GenBank mRNA | AB099081.1 | - | 2 - 3 | 87 | 80 | |
| AY528251.1 | - | 1 - 2 | 60 | 80 | ||
| AY911345.1 | - | 2 - 3 | 103 | 80 | ||
| BC102500.1 | - | 1 | 104 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102500.1, NM_001014387.2 transcripts.
Set Membership: |
|

RPS12
Genomic Map