Assay Details
| Gene Symbol: RPS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014387.2 | NP_001014409.1 | 2 - 3 | 171 | 66 | |
| GenBank mRNA | AB098828.1 | - | 3 - 4 | 367 | 66 | |
| AB099081.1 | - | 3 - 4 | 200 | 66 | ||
| AY528251.1 | - | 2 - 3 | 173 | 66 | ||
| AY911345.1 | - | 3 - 4 | 216 | 66 | ||
| BC102500.1 | - | 1 | 217 | 66 | ||
| DQ347612.1 | - | 3 - 4 | 217 | 66 | ||
More Information
Set Membership: |
|

RPS12
Genomic Map