Assay Details
| Gene Symbol: RPS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014387.2 | NP_001014409.1 | 4 - 5 | 370 | 63 | |
| GenBank mRNA | AB099081.1 | - | 5 - 6 | 399 | 63 | |
| AY528251.1 | - | 4 - 5 | 372 | 63 | ||
| AY911345.1 | - | 5 - 6 | 415 | 63 | ||
| BC102500.1 | - | 1 | 416 | 63 | ||
More Information
Set Membership: |
|

RPS12
Genomic Map