Assay Details
| Gene Symbol: AGPAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015537.1 | NP_001015537.1 | 7 - 8 | 1125 | 63 | |
| GenBank mRNA | BT020709.1 | - | 7 - 8 | 1125 | 63 | |
| BT020858.1 | - | 7 - 8 | 1086 | 63 | ||
More Information
Set Membership: |
|

AGPAT4
Genomic Map